Anti-PTTG2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP16911C
89512-336EA 642.9 USD
89512-336
Anti-PTTG2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000, Immunofluorescence: 1:10~50,

Type: Primary
Antigen: PTTG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR