Anti-ERBB2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP3781G
89512-296EA 646.72 USD
89512-296
Anti-ERBB2 Rabbit Polyclonal Antibody
Antibodies
Immunofluorescence: 1:100

Type: Primary
Antigen: ERBB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR