Anti-RUNX3 Rabbit Polyclonal Antibody

Supplier: Abgent
AP14667C
89512-282EA 642.73 USD
89512-282
Anti-RUNX3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000, Immunhistochemsitry: 1:10~50, Immunofluorescence: 1:10~50,

Type: Primary
Antigen: RUNX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR