Anti-CD55 Rabbit Polyclonal Antibody

Supplier: Abgent
AP14798A
89512-278EA 646.72 USD
89512-278
Anti-CD55 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000, Immunhistochemsitry: 1:10~50, Flow Cytometry: 1:10~50, Immunofluorescence: 1:10~50,

Type: Primary
Antigen: CD55
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR