Anti-CTNNB1 Rabbit Polyclonal Antibody

Supplier: Abgent
AX10009
89512-260EA 646.4 USD
89512-260
Anti-CTNNB1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:2000, Immunhistochemsitry: 1:50, Flow Cytometry: 1:50, Immunofluorescence: 1:50,

Type: Primary
Antigen: CTNNB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR