Anti-TNNI3 Mouse Monoclonal Antibody [clone: 358CT10.3.6]

Supplier: Abgent
AM1979A
89512-236EA 646.72 USD
89512-236
Anti-TNNI3 Mouse Monoclonal Antibody [clone: 358CT10.3.6]
Antibodies
Western Blot: 1:500~8000,

Type: Primary
Antigen: TNNI3
Clonality: Monoclonal
Clone: 358CT10.3.6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR