Anti-CAMK2A Mouse Monoclonal Antibody [clone: 390CT12.4.4]

Supplier: Abgent
AM2002A
89512-220EA 646.72 USD
89512-220
Anti-CAMK2A Mouse Monoclonal Antibody [clone: 390CT12.4.4]
Antibodies
LEAF (Low Endotoxin, Azide-Free)

Western Blot: 1:1000~8000,

Type: Primary
Antigen: CAMK2A
Clonality: Monoclonal
Clone: 390CT12.4.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR