Anti-FABP3 Mouse Monoclonal Antibody [clone: 388CT12.2.4.1]

Supplier: Abgent
AM2000A
89512-216EA 646.72 USD
89512-216
Anti-FABP3 Mouse Monoclonal Antibody [clone: 388CT12.2.4.1]
Antibodies
Western Blot: 1:1000~4000,

Type: Primary
Antigen: FABP3
Clonality: Monoclonal
Clone: 388CT12.2.4.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR