Anti-GJC2 Mouse Monoclonal Antibody [clone: 391CT6.4.3]

Supplier: Abgent
AM1998A
89512-214EA 646.72 USD
89512-214
Anti-GJC2 Mouse Monoclonal Antibody [clone: 391CT6.4.3]
Antibodies
Western Blot: 1:100~8000,

Type: Primary
Antigen: GJC2
Clonality: Monoclonal
Clone: 391CT6.4.3
Conjugation: Unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR