Anti-ESR1 Mouse Monoclonal Antibody [clone: 415CT16.3.2.1]

Supplier: Abgent
AM2006A
89512-166EA 615.93 USD
89512-166
Anti-ESR1 Mouse Monoclonal Antibody [clone: 415CT16.3.2.1]
Antibodies
Western Blot: 1:500~3200,

Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: 415CT16.3.2.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR