Anti-B2M Mouse Monoclonal Antibody [clone: 467CT12.3.1]

Supplier: Abgent
AM2052A
89512-114EA 646.72 USD
89512-114
Anti-B2M Mouse Monoclonal Antibody [clone: 467CT12.3.1]
Antibodies
Western Blot: 1:1000~16000,

Type: Primary
Antigen: B2M
Clonality: Monoclonal
Clone: 467CT12.3.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR