Anti-PDK3 Mouse Monoclonal Antibody [clone: 431CT6.1]

Supplier: Abgent
AM2038A
89512-096EA 646.55 USD
89512-096
Anti-PDK3 Mouse Monoclonal Antibody [clone: 431CT6.1]
Antibodies
Western Blot: 1:100~800,

Type: Primary
Antigen: PDK3
Clonality: Monoclonal
Clone: 431CT6.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR