Anti-SPRED1 Mouse Monoclonal Antibody [clone: 462CT2.5.1]

Supplier: Abgent
AM2036A
89512-092EA 615.93 USD
89512-092
Anti-SPRED1 Mouse Monoclonal Antibody [clone: 462CT2.5.1]
Antibodies
Western Blot: 1:100~1600,

Type: Primary
Antigen: SPRED1
Clonality: Monoclonal
Clone: 462CT2.5.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR