Anti-FKRP Mouse Monoclonal Antibody [clone: 386CT13.1.9]

Supplier: Abgent
AM2033A
89512-086EA 615.93 USD
89512-086
Anti-FKRP Mouse Monoclonal Antibody [clone: 386CT13.1.9]
Antibodies
Western Blot: 1:100~400,

Type: Primary
Antigen: FKRP
Clonality: Monoclonal
Clone: 386CT13.1.9
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR