Anti-DDIT3 Mouse Monoclonal Antibody [clone: 563CT2.4.1]

Supplier: Abgent
AM2156A
89512-054EA 646.72 USD
89512-054
Anti-DDIT3 Mouse Monoclonal Antibody [clone: 563CT2.4.1]
Antibodies
Western Blot: 1:500~1600,

Type: Primary
Antigen: DDIT3
Clonality: Monoclonal
Clone: 563CT2.4.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR