Anti-MVP Mouse Monoclonal Antibody [clone: 599CT2.4.1]

Supplier: Abgent
AM2146A
89512-042EA 646.72 USD
89512-042
Anti-MVP Mouse Monoclonal Antibody [clone: 599CT2.4.1]
Antibodies
Western Blot: 1:100~1600,

Type: Primary
Antigen: MVP
Clonality: Monoclonal
Clone: 599CT2.4.1
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR