Anti-VSIG8 Mouse Monoclonal Antibody [clone: 604CT14.1.2]

Supplier: Abgent
AM2124A
89512-020EA 646.72 USD
89512-020
Anti-VSIG8 Mouse Monoclonal Antibody [clone: 604CT14.1.2]
Antibodies
Western Blot: 1:100~800,

Type: Primary
Antigen: VSIG8
Clonality: Monoclonal
Clone: 604CT14.1.2
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgM
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR