Anti-FOXA1 Mouse Monoclonal Antibody [clone: 596CT18.4.3]

Supplier: Abgent
AM2103A
89511-988EA 646.72 USD
89511-988
Anti-FOXA1 Mouse Monoclonal Antibody [clone: 596CT18.4.3]
Antibodies
Western Blot: 1:100~200,

Type: Primary
Antigen: FOXA1
Clonality: Monoclonal
Clone: 596CT18.4.3
Conjugation: Unconjugated
Epitope: internal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR