Dispenser for Parafilm® Sealing Film
Supplier: Heathrow Scientific
HS234525C HS234525D
89511-852EA 85.54 USD
89511-852 89511-854
Dispenser for Parafilm® Sealing Film
Sealing Film Dispensers
ABS plastic dispenser designed with integrated safety razor that slides smoothly creating a clean cut.
  • Translucent dispenser allows visibility to roll supply
  • Storage bin on back of the dispenser keeps pens and other small accessories handy
Dispenser holds one or two 2" (50 mm) rolls or one 4" (100 mm) roll.
Store, dispense, and cut Parafilm® M tape, label rolls including Tough-Tags®.
Delivery information: Supplied with two safety blades.