VWR® Personal Low Temperature Incubator
Supplier: VWR International
50142296
89511-416EA 966.45 USD
89511-416
VWR® Personal Low Temperature Incubator
Incubators Cooled Incubators
Save space with a VWR® personal low temperature incubator ideal for small sample work.
  • Space-saving design in 0.64 cu.ft. volume
  • Peltier technology with fan for optimal temperature performance
  • Temperature range 17°C - 40°C
  • Monitor samples through transparent door with internal light
  • Easy-to-use control panel
  • Digital display for easy reading
  • Two perforated stainless steel shelves included
  • 2-year warranty
Common applications include microbiology, blood sample storage, bacterial research, histology, coliform determination, yeast growth.
Certifications: CE.