Slide Mailer, Screw-Cap, Globe Scientific
Supplier: Globe Scientific
513068
89510-784CS 483.5 USD
89510-784
Slide Mailer, Screw-Cap, Globe Scientific
Slide Mailers
This screw-cap slide mailer is produced from Polypropylene (PP) and can hold 4 standard sized microscope slides (25 x 755 mm or 3 x 1")
The inner slots keep slides separated to avoid cross-contamination during shipping or storage.