Slide Dispenser for 72 Slides, Globe Scientific
Supplier: Globe Scientific
513000
89510-754EA 50.57 USD
89510-754
Slide Dispenser for 72 Slides, Globe Scientific
Slide Holders
The Globe microscope slide dispenser is designed for use with 25 x 75 mm and 3 x 1" microscope slides
To use, simply insert one full box of 72 slides into the holder. Turn the dial to dispense one slide at a time. This convenient dispenser prevents slides from getting dirty, dusty and scratched.