ALCH 50%, Reagent Grade for histology (RGA 50)

Supplier: VWR International
Danger

89508-646
89508-646CS 140.14 USD
89508-646
ALCH 50%, Reagent Grade for histology (RGA 50)
Alcohol

Our reagent grade alcohol is a denatured form of ethyl alcohol, approved for sale under U.S. regulations (90 parts ethyl alcohol, 5 parts methyl alcohol, and 5 parts isopropyl alcohol)


  • Preferred mixture for dehydrating tissue samples prior to embedding

MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1987
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR