Epower™ Penicillium aurantiogriseum, Microbiologics
Supplier: MICROBIOLOGICS, INC.
0794E3
89506-234EA 462.07 USD
89506-234
Epower™ Penicillium aurantiogriseum, Microbiologics
Controls and Calibration Verification Materials
Epower™ QC microorganisms can be used to perform many essential quality control functions including microbial detection and enumeration, equipment calibration, method validation, bioburden determination, and antibacterial effectiveness and lethality testing
Epower™ is packaged in a resealable vial containing 10 pellets of a single quantitative QC microorganism strain. An online certificate of analysis containing documentation of phenotypic, macroscopic and microscopic test results is available.
Available in concentrations ranging from 102 to 108 CFU per pellet, Epower™ lyophilized microorganism preparations are extremely versatile. Epower™ pellets can be used individually or several Epower™ pellets can be combined for a mixed microorganism population challenge. They can also be easily manipulated to deliver a wide variety of concentrations. For added convenience, a peel-off Certificate of Assay is included for quality control documentation.
Certifications: Assured quality, meets USP guidelines.
Ordering information: To receive this product, customers must have a valid ATCC End User Licensing Agreement in place with Microbiologics. Registration for this agreement is only required once, per location during the first order. Instructions for how to register can be downloaded from the website. Go to the "Related Information" box, click on "Additional Product Details", then "Instructions for End User Registration with Microbiologics".
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.