LYFO DISK Staphylococcus aureus subsp. aureus ATCC® 29213™, Microbiologics
Supplier: MICROBIOLOGICS, INC.
0365L
89503-862EA 182.15 USD
89503-862
LYFO DISK Staphylococcus aureus subsp. aureus ATCC® 29213™, Microbiologics
Controls and Calibration Verification Materials
LYFO DISK® is a lyophilized pellet containing a single strain of a QC microorganism and is the most economical reference stock culture option
Weak beta-lactamase producing strain, EUCAST control for antimicrobial susceptibility testing (disk diffusion and MIC determination), CLSI control for antimicrobial susceptibility testing (MIC determination) oxacillin sensitive mecA negative.
LYFO DISK® is known for its ease-of-use; simply re-hydrate the pellet and inoculate. LYFO DISK® is available in a re-sealable vial containing 6 pellets of a single QC microorganism strain.
Certifications: LYFO DISK® falls under the category of In-Vitro Diagnostics Medical Devices Directives for CE Mark Conformity. This product is also listed with the FDA.
Ordering information: To receive this product, customers must have a valid ATCC End User Licensing Agreement in place with Microbiologics. Registration for this agreement is only required once, per location during the first order. Instructions for how to register can be downloaded from the website. Go to the "Related Information" box, click on "Additional Product Details", then "Instructions for End User Registration with Microbiologics".
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.