Letheen Agar Modified, with Tween 80 1.5%, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
W55
89501-560PK
66.17
USD
89501-560
Letheen Agar Modified, with Tween 80 1.5%, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
For the cultivation and enumeration of microorganisms from cosmetics and personal care product samples.
For the cultivation and enumeration of microorganisms from cosmetics and personal care product samples.
Ordering information: Minimum order of 10 packs