SP4 Broth with Glucose, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
U86
89501-558EA 200.02 USD
89501-558
SP4 Broth with Glucose, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
SP4 Media are recommended for the isolation, differentiation and maintenance of mycoplasma, including M. hominis, M. pneumoniae, and Ureaplasma urealyticum.
Ordering information: Minimum order of 10
Caution: Upon receipt store at 2-8°C away from direct light.