CulGenex™ LB Broth, Lennox, Dehydrated Culture Media, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
C7659
89501-486PK 454.12 USD
89501-486
CulGenex™ LB Broth, Lennox, Dehydrated Culture Media, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
recommended for maintaining and cultivating recombinant strains of Escherichia coli in molecular biology procedures.
Hardy Diagnostics CulGenex™ LB Broth, Lennox is recommended for maintaining and cultivating recombinant strains of Escherichia coli for molecular biology procedures.
Hardy Diagnostics CulGenex™ Dehydrated Culture Media is formulated to meet or exceed the highest standards. Each lot of CulGenex™ media is meticulously tested for performance and reproducibility. Over 30 years of formulating and media manufacturing experience.
Container sizes available:
2L mylar pouch – pre-measured for a 2 liter batch, no weighing needed!
500ml mylar pouch - pre-measured for a 500ml batch, 6 pouches per pack
500gm bottle - wide mouth, easy grip bottle, comes with a free scoop!
2kg and 10kg bucket - with convenient screw lid