VWR® LB Broth Liquid
Supplier: VWR
J833-1L
89500-596EA
64.44
USD
89500-596
VWR® LB Broth Liquid
Media for Microbiology
Prepared Media for Microbiology
LB Broth, Liquid is used for the propagation and maintenance of E.coli. Widely used for the preparation of plasmid DNA and recombinant proteins.
- Ready to use liquid media
- Clear, yellow to brown liquid
1L of media contains 10 grams of sodium chloride, 10 grams of tryptone/ cesein peptone, 5 grams of yeast extract bacteriological.
No growth detected in Bioburden (Milliflex) (EQC) test. pH @25°C is 6.65 - 7.15. It has refractive index of 1.334 - 1.342. Specific Gravity at 20°C is 1.0116 - 1.0128. Abs.@260nm (1:100, Water) is 0.2263 - 0.2663.