VWR® Ypd Media Liquid
Supplier: VWR
1B1493-1L
89500-592EA 64.87 USD
89500-592
VWR® Ypd Media Liquid
Media for Microbiology Prepared Media for Microbiology
YPD Media, Liquid is a complete medium for the propagation and maintenance of yeast
The composition of YPD is (w/v) 1% yeast extract, 2% peptone and 2% D-glucose.