Triethylamine, high purity

Supplier: VWR
Danger

Synonyms: (Diethylamino)ethane, TEA

1B1180-100ML
89500-556CS 639.46 USD
89500-556 89500-558
Triethylamine, high purity
Triethylamine
Formula: (C₂H₅)₃N
Boiling Pt: 90 °C (1013 hPa)
Melting Pt: –115 °C
Density: 0.729 g/cm³ (20 °C)
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00009051
CAS Number: 121-44-8
EINECS: 204-469-4
UN: 1296
ADR: 3,II
Merck Index: 13,09740
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2b1f184e438ecb01f88a4d90ee46c0d4316859d&vid=1c68408a0eedf1bfdb02d1b1e2337d50d449cf9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Color (APHA) 15
Diethylamine 0.2 %
Ethanol 0.2 %
Ethylamine 0.2 %
Moisture (KF) 0.2 %
N-Ethylidene-Ethylamine 0.1 %
Purity 99.5 %
Specific Gravity 0.726 - 0.73


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR