Plain Microscope Slides, Globe Scientific
Supplier: Globe Scientific
1321 1301 1331
89500-466CS 570.92 USD
89500-466 89500-468 10118-594
Plain Microscope Slides, Globe Scientific
Slides Microscope Slides
Globe Scientific microscope slides are produced from premium glass sheets and offer exceptional quality while providing considerable savings.
  • Dimensions: 25×75×1 mm
  • Thickness: 1.1 mm
Globe Scientific plain microscope slides are available in three styles. Choose from ground edge, ground edge with safety corners (slightly clipped corners for comfort), or beveled edge with clipped corners. The slides are pre-cleaned and supplied ready for use.
Ordering information: Slides are packaged 72 per box, 20 boxes per case (10 gross).