VWR® Traceable® Digital Humidity Temperature/Dew Point Meter
Supplier: VWR International
93678-26 VC4800
89500-398EA 143.04 USD
89500-398 10754-538 10754-724
VWR® Traceable® Digital Humidity Temperature/Dew Point Meter
Hygrometers Digital Thermo-Hygrometers
Convenient triple memories recall minimum/maximum readouts for relative humidity, dew point, and temperature.
  • Simultaneously displays all three current readings
  • Sits anywhere in the lab and updates every 30 seconds
  • Relative humidity range: 1 to 99%
  • Resolution: 0.1%
  • Accuracy: ±3% mid-range, ±5% elsewhere
  • Resolution: 0.1°
Simultaneously displays all three current readings. Always-on feature allows for monitoring conditions all night, over weekends, and for extended time periods. Audible and visual LED alarm may be set to trigger at three different humidity readings (55%, 60% or 65%). Solid state sensors deliver response times of less than 30 seconds. Oversized display supplies updates once a second.