ABsolute™ QPCR Seal Roll, Thermo Scientific
Supplier: Thermo Fisher Scientific
AB-2170
89499-574CS 9222.82 USD
89499-574
ABsolute™ QPCR Seal Roll, Thermo Scientific
Seals Microplate Seals
Clear adhesive seal prevents evaporation during sample storage or thermal cycling, while still allowing samples to be accessed by piercing foil with pipette tips.
  • Optically clear, peelable, no residue
  • For fluorescence applications such as QPCR
  • PCR Film seals are recommended for use in conjunction with a cycler that has a screw-down or clip-down lid