Whatman™ Multicompressor Tray for Mini UniPrep G2 Syringeless HPLC Filter
Supplier: Cytiva
MUPG2MCWT8
89499-528EA 33.6 USD
89499-528
Whatman™ Multicompressor Tray for Mini UniPrep G2 Syringeless HPLC Filter
Presses
A Mini UniPrep G2 multicompressor tray allows you to preload eight samples, further increasing the time savings provided by Cytiva's Mini-UniPrep G2 syringeless filtration line.
  • A multicompressor streamlines syringeless filtration - preload in advance with an extra tray
  • Designed for Cytiva's Mini-UniPrep G2, which combines a borosilicate glass vial, plunger and attached filter membrane, and septum with cap
The borosilicate glass design of the Mini-UniPrep G2 minimizes the risk of leachables contaminating a sample, making it an excellent choice for pharmaceutical quality control (pharma QC) and water monitoring applications.
Browse the Whatman line from Cytiva's business to find the right syringeless HPLC filter and compressor for your application.
Whatman Mini UniPrep G2 can be used with a multicompressor that can process up to eight samples at one time. An extra tray allows you to prepare samples in advance, resulting in reduced overall filtration time.
The glass design prevents interference from hydrocarbon extractables that could be found in plastic.