Microcentrifuge Storage Box, Simport Scientific
Supplier: Simport Scientific
T330-64
89499-338CS 590.8 USD
89499-338
Microcentrifuge Storage Box, Simport Scientific
Boxes Boxes for Tubes
This Microcentrifuge Storage Box with a polyurethane foam insert is ideal for holding up to 64 tubes from 0.5 mL to 2 mL
Can withstand temperature range of -90 to 80°C. A transparent cover allows you to see the contents of the box, and is keyed to the base in order to prevent misalignment. Stackable. Made of high impact polystyrene.