Gastight® CTC PAL® S-Line Syringes for HPLC Autosamplers, Hamilton
Supplier: Hamilton
67444-01 67446-01 67450-01 67452-01 67442-01 67448-01
89498-972EA 120.68 USD
89498-972 89498-974 89498-976 89498-978 89498-980 89498-982
Gastight® CTC PAL® S-Line Syringes for HPLC Autosamplers, Hamilton
Syringes Syringes for Chromatography
CTC PAL autosamplers are one of the most popular sampling devices on the market today
S-Line syringes are the cost-effective choice for CTC PAL autosampler applications. S-Line syringes deliver great performance in everyday use for HPLC injections at an exceptional value. The Gastight® S-Line syringe is an ideal choice for HPLC sample injections. It incorporates a precision-machined PTFE plunger tip which creates a leak-free seal. With the tight fit in the glass barrel, the tip wipes the interior of the syringe barrel free of sample, minimizing sample carryover. This feature is particularly useful with heterogeneous samples as it reduces the chance that a deposit will occur and cause the plunger to freeze.
Customers rely on their ability to sample from many different vial and well types and on their ultimate platform flexibility. The syringe is the heart of every injection that the HPLC PAL system performs. The latest generation of Hamilton autosampler syringes is recommended by CTC for use in the popular HPLC PAL systems: PAL HTC = xt, PAL HTS = xt, PAL HTX = xt.
Certifications: UL Listed. CE compliant.
Ordering information: For questions about Hamilton CTC syringes and compatability with your CTC PAL autosampler, please contact your Avantor representative.