Microliter™ and Gastight® CTC PAL® GC Autosampler Syringes, S-Line Syringes, Hamilton
Supplier: Hamilton
67436-01 67454-01 67438-01 67440-01 67430-01 67434-01
89498-960EA 113.74 USD
89498-960 89498-962 89498-964 89498-966 89498-968 89498-970
Microliter™ and Gastight® CTC PAL® GC Autosampler Syringes, S-Line Syringes, Hamilton
Syringes Syringes for Chromatography
CTC PAL autosamplers are one of the most popular sampling devices on the market today
S-Line syringes are the cost-effective choice for CTC PAL autosampler applications. S-Line syringes deliver great performance in everyday use for GC injections at an exceptional value. The Gastight® version of the S-Line syringe is an ideal choice for GC sample injections. It incorporates a precision-machined PTFE plunger tip which creates a leak-free seal. With the tight fit in the glass barrel, the tip wipes the interior of the syringe barrel free of sample, minimizing sample carryover. This feature is particularly useful with heterogeneous samples as it reduces the chance that a deposit will occur and cause the plunger to freeze.
Customers rely on their ability to sample from many different vial and well types and on their ultimate platform flexibility. The syringe is the heart of every injection that the GC PAL system performs. The latest generation of Hamilton autosampler syringes is recommended by CTC for use in the popular GC PAL systems: PAL HTC = xt, PAL HTS = xt, PAL HTX = xt.
Certifications: UL Listed. CE compliant.
Ordering information: Accessories not included. For questions about Hamilton CTC syringes and compatability with your CTC PAL autosampler, please contact your Avantor representative.