Samco™ Autoanalyzer Sample Cups, Thermo Scientific
Supplier: Avantor
89498-492CS 7738.07 USD
89498-492 89498-494 89498-506 89498-508 89498-510
Samco™ Autoanalyzer Sample Cups, Thermo Scientific
Sample Containers Sample Cups for Analyzers
A range of Samco™ autoanalyzer cups suitable for use in a wide range of analyzers.
  • High clarity polystyrene
  • Precision molded graduations for accurate volume handling
  • Resistant to most aqueous solutions
Certifications: FDA Class 1 Exempt Medical Device.
Ordering information: Ambient storage and shipping recommended for containers.