Samco™ Bio-Tite™ Specimen Container, 40 ml/48 mm (1 oz.), Thermo Scientific
Supplier: Samco
04 0181 04 0506
89498-390CS 405.62 USD
89498-390 89498-396
Samco™ Bio-Tite™ Specimen Container, 40 ml/48 mm (1 oz.), Thermo Scientific
Sample Containers General Purpose Containers
Prevent bio-hazardous spills with the leak-resistant Samco™ Bio-Tite™ Wide-Mouth 40mL (1oz.) 48mm Specimen Containers
  • Plug seal cap ensures excellent leak-resiatant performance
  • Cap and base knurls ensure ease of handling
  • Supplied nonsterile
These stackable, shatter-resistant containers are manufactured from virgin polypropylene and are ideal for the collection and transportation of both liquid and solid samples. Supplied bulk packed, containers are ideal for pre-fill applications.
Ordering information: Ambient storage and shipping recommended for all containers.