Samco™ Wide Mouth Bio-Tite™ Specimen Containers, 90 ml, Thermo Scientific
Supplier: Samco
02 1106 02 1016 02 1038 02 0180 02 1051 02 1102 02 1104 02 1105 02 1140 02 1181
14670-478CS 538.14 USD
14670-478 14670-476 95043-674 89498-330 89498-346 89498-348 89498-350 89498-352 89498-354 89498-358
Samco™ Wide Mouth Bio-Tite™ Specimen Containers, 90 ml, Thermo Scientific
Sample Containers General Purpose Containers
Prevent bio-hazardous spills while conserving laboratory space with stackable Thermo Scientific™ Samco™ Narrow-Mouth Bio-Tite™ 90 mL (3 oz.), 53 mm specimen containers
These shatter-resistant containers feature a proprietary, all plastic, leak-resistant seal. They are tested at varying pressures, making them ideal for use in various transport systems. Containers are available pre-assembled with caps attached or bagged separately.