Samco™ Bio-Tite™ Specimen Container, Narrow Mouth, 90 ml/48 mm (3 oz.), Thermo Scientific
Supplier: Samco
02 0147 02 0516
89498-326CS 341.83 USD
89498-326 89498-340
Samco™ Bio-Tite™ Specimen Container, Narrow Mouth, 90 ml/48 mm (3 oz.), Thermo Scientific
Sample Containers General Purpose Containers
Prevent bio-hazardous spills while conserving laboratory space with stackable Samco™ Bio-Tite™ Narrow-Mouth 90mL (3oz.) 48mm Specimen Containers
  • Plug seal cap ensures excellent leak-resiatant performance
  • Cap and base knurls ensure ease of handling
  • Containers are stackable on top of each other for ease of storage
  • Supplied nonsterile or gamma irradiated for guaranteed sterility
These shatter-resistant containers feature a leak-resistant seal and are ideal for the collection and transport of both liquid and solid samples. Available in both sterile and nonsterile formats, with a range of cap colors and label types to suit most applications.
Certifications: FDA Class 1 Exempt Medical Device.
Ordering information: Ambient storage and shipping recommended for all containers.