Wall Mounted Drying Rack, Globe Scientific
Supplier: Globe Scientific
600213
89498-072EA 601.18 USD
89498-072
Wall Mounted Drying Rack, Globe Scientific
Racks Draining Racks
This 72-place wall mountable laboratory drying rack is ideal for draining and drying all types of Labware
The back plate of the support is made from a single mold of 4 mm rigid high-impact polystyrene (HIPS). The peg sockets feature closed ends, eliminating leakage and consequent biohazards. The seamless drip channel collects waste which drains via a supplied drain tube.
The 72 (push in) pegs can accommodate items with a 15 mm+ neck bore. They are easily removable for cleaning or to accommodate oddly shaped items. Also included are 11 small pegs to accommodate small diameter items with a neck bore of 6mm+ (such as test tubes). This durable rack will not rust or stain.
The rack is supplied with a mounting kit to facilitate convenient hanging.