Centrifuge Tube Rack, 15 and 50 ml Globe Scientific

Supplier: Globe Scientific
456930
89498-062EA 34.91 USD
89498-062
Centrifuge Tube Rack, 15 and 50 ml Globe Scientific
Racks Tube Racks
Rack is designed to hold both 15 and 50 ml centrifuge tubes simultaneously.

  • Dimensions: 209×171×63 mm (8.2×6.7×2.4")
  • Material: acrylonitrile butadiene styrene (ABS)

Ships flat and is simple to assemble. The rack accomodates 20×50 ml tubes and 30×15 ml tubes.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR