VWR® Nitrogen Generators
Supplier: VWR International
89497-476
89497-476EA 57517.89 USD
89497-476
VWR® Nitrogen Generators
Gas Generators Nitrogen Generators
The VWR® LC/MS NitroFlowLab is a self contained generator that produces up to 60 slpm of pure LC/MS grade nitrogen at pressures of up to 110 psig.
  • Complete plug and play system recommended for all major LC/MS instruments
  • Phthalate-free, no organic vapors
  • Eliminate dangerous nitrogen cylinders from the laboratory
  • Nearly silent operation, operates at less than 49 dB(A)
Nitrogen is produced utilizing a combination of a scroll compressor and nitrogen membrane separation technologies
Typical applications include LC/MS, nebulizer gases for APCI and ESI, Jet Stream, I Funnel, ELSD, Turbo Vaps and chemical solvent evaporation.
This combination of technologies yields the highest performing, most reliable and quietest integrated nitrogen generation system available.