VWR® Heat Resistant Film for Thermal Cycling
Supplier: VWR International
89496-565
89496-565CS 113.34 USD
89496-565
VWR® Heat Resistant Film for Thermal Cycling
Seals Microplate Seals
VWR® Heat Resistant Films are improved films for classic thermal cycling applications.
  • Improved adhesion
  • Reduced evaporation
  • Longer end tabs for easier handling
  • Heat-resistant
They are made of 50 Uu Polyester with a 41 Um layer of acrylic adhesive
VWR® Heat Resistant Films are recommended for temperatures from –40 to 120°C. Films are certified DNase-, RNase-, and nucleic acid-free.
The films offer better adhesion and reduced evaporation. They feature longer end tabs for easier handling.