VWR® Incubator Verification Thermometer, Traceable to NIST
Supplier: VWR International
60200-0911
89495-974CS 300.55 USD
89495-974
VWR® Incubator Verification Thermometer, Traceable to NIST
Thermometers Analog Thermometers
These thermometers are certified at one specific temperature.
  • PFA coating
  • Mercury-free
  • Permanently fused markings
  • Lead-free glass
PFA coated for enhanced safety in case of breakage, they are used to verify sample temperatures in incubators covering ranges from 15 to 50°C
Ideal for the chemical, petro-chemical, pharmaceutical, medical, cosmetic and dairy industries, these thermometers are found in universities, hot houses, scientific institutes, and incubator/oven/laboratory equipment producing facilities.
They contain EnviroKleen™ certified Enviro-Safe® liquid and feature permanently fused markings and lead free glass. Insertion of the thermometer into the included recyclable plastic bottle, filled with EnviroKleen™ certified Enviro-Safe® liquid, protects samples from contamination by eliminating the need for direct contact. Bottle contents mimic the sample for more accurate temperature monitoring. Enviro-Safe® thermometer fill/Enviro-Safe® bottle fill.
Certifications: Manufactured and tested by Avantor's USA quality team, these verification thermometers are traceable to NIST and include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability. Individually serialized.
Delivery information: Each thermometer is supplied with a NIST Statement of Accuracy and Traceability.