VWR® Freezer Verification Thermometer with Certificate of Compliance, Traceable to NIST
Supplier: VWR International
60200-0711
89495-970CS 300.26 USD
89495-970
VWR® Freezer Verification Thermometer with Certificate of Compliance, Traceable to NIST
Thermometers Analog Thermometers
These thermometers are certified at one specific temperature.
  • PFA coating
  • Mercury-free
  • Permanently fused markings
  • Lead-free glass
PFA coated for enhanced safety in case of breakage, they are used to verify sample temperatures in freezers covering ranges from -30 to 1°C
Ideal for the chemical, petro-chemical, pharmaceutical, medical, cosmetic, and dairy industries, these thermometers are found in universities, breweries, refrigeration houses, scientific institutes, blood banks, freezer and laboratory equipment producing facilities.
These mercury-free thermometers are filled with EnviroKleen™ certified Enviro-Safe® liquid, and have permanently fused markings and lead-free glass. Insertion of the thermometer into the included recyclable plastic bottle, filled with glass beads or EnviroKleen™ certified Enviro-Safe® liquid, protects samples from contamination by eliminating the need for direct contact. Bottle contents mimic the sample for more accurate temperature monitoring. Enviro-Safe® thermometer fill/Enviro-Safe® bottle fill.
Certifications: Manufactured and tested by Avantor's USA quality team, these verification thermometers are traceable to NIST and include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability. Individually serialized.