C-MAG MS 10 IKAMAG® Magnetic Stirrer, 230 V
Supplier: IKA Works
3582600
89495-334EA 1289.26 USD
89495-334
C-MAG MS 10 IKAMAG® Magnetic Stirrer, 230 V
Stirrers Magnetic Stirrers
New magnetic stirrer without heating features a set-up plate which offers excellent chemical resistance.
  • 230 V model
Units are equipped with a powerful motor for stirring quantities of up to 15 L (H₂O). The elevated control panel is designed for protection against liquid leaks.
Units meet IP 21 protection class according to DIN EN 60529.
Certifications: CE marked. UL and C-UL listed.
Ordering information: Supplied with a three-year manufacturer's warranty (2+1 with product registration at www.ika.com/register). For additional laboratory accessory options, see the IKA® Lablift Manual (see 89495-302 series). Accessories are sold separately.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (89495-332) is also available, please see separate product page.