C-MAG HS 10 Digital IKAMAG® Hot Plate Stirrer, 230 V
Supplier: IKA Works
4240400
89495-314EA 1763.89 USD
89495-314
C-MAG HS 10 Digital IKAMAG® Hot Plate Stirrer, 230 V
Stirrers Magnetic Hotplate Stirrers
New magnetic stirrer features heating capabilities and a ceramic heating plate that offers excellent chemical resistance.
  • 230 V model
Units are equipped with a powerful motor for stirring quantities of up to 15 L (H₂O). The LCD display provides simultaneous digital display of target and actual temperatures. Direct connection for a PT 1000 temperature sensor enables precise temperature control (included in delivery). Control accuracy in medium ±0.5 K (in combination with PT 1000). Display of actual temperature value in medium with 0.1 K resolution when using PT 1000 temperature sensor. Units offer three operating modes to choose from (Standard, Safe, Adjustment Protection). Standard safety features include, a fixed safety circuit of 550 °C, a Hot Top indicator for hot surface warning to prevent burns, a digital error code display, and an elevated control panel for protection against leaks.
Units meet IP 21 protection class according to DIN EN 60529.
Certifications: CE marked.
Ordering information: Supplied with three-year manufacturer's warranty (2+1 with product registration at www.ika.com/register). PT 1000 temperature sensor is included in delivery. For additional laboratory accessory options, see the IKA® Lablift Manual (see 89495-302 series). Other accessories are sold separately.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (89495-312) is also available, please see separate product page.